Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2-Methylbutyric acid, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
116-53-0
C5H10O2
102.133
MFCD00002669
WLAMNBDJUVNPJU-UHFFFAOYSA-N
2-methylbutyric acid,dl-2-methylbutyric acid,butanoic acid, 2-methyl,ethylmethylacetic acid,methylbutyric acid,2-methyl butyric acid,2-methybutyric acid,methylethylacetic acid,active valeric acid,butyric acid, 2-methyl
8314
CHEBI:37070
2-methylbutanoic acid
CCC(C)C(=O)O
116-53-0
C5H10O2
102.133
MFCD00002669
WLAMNBDJUVNPJU-UHFFFAOYSA-N
2-methylbutyric acid,dl-2-methylbutyric acid,butanoic acid, 2-methyl,ethylmethylacetic acid,methylbutyric acid,2-methyl butyric acid,2-methybutyric acid,methylethylacetic acid,active valeric acid,butyric acid, 2-methyl
8314
CHEBI:37070
2-methylbutanoic acid
CCC(C)C(=O)O
Refractive Index 1.4045 to 1.4065 (20°C, 589nm)
Packaging Glass bottle
Density 0.936g/mL
Infrared Spectrum Authentic
Quantity 100g
Color Colorless to Yellow
Assay Percent Range 98%
Linear Formula C2H5CH(CH3)CO2H
Beilstein 02, 305
Formula Weight 102.13
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material DL-2-Methylbutyric acid, 98%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-982-7