Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2-Methylglutamic Acid Hemihydrate 98.0+%, TCI America™

Click to view available options
Quantity:
100mg
1g
71-90-9
C6H11NO4
161.157
MFCD00004188
QHSCIWIRXWFIGH-UHFFFAOYSA-N
dl-2-methylglutamic acid,2-methylglutamic acid,dl-glutamic acid, 2-methyl,2-amino-2-methylglutaric acid,alpha-methylglutamic acid,glutamic acid, 2-methyl,hemi-hydrate,alpha-methylglutamate,2-methylglutamic acid, dl,a-methylglutamic acid
95440
2-amino-2-methylpentanedioic acid
CC(CCC(=O)O)(C(=O)O)N
71-90-9
C6H11NO4
161.157
MFCD00004188
QHSCIWIRXWFIGH-UHFFFAOYSA-N
dl-2-methylglutamic acid,2-methylglutamic acid,dl-glutamic acid, 2-methyl,2-amino-2-methylglutaric acid,alpha-methylglutamic acid,glutamic acid, 2-methyl,hemi-hydrate,alpha-methylglutamate,2-methylglutamic acid, dl,a-methylglutamic acid
95440
2-amino-2-methylpentanedioic acid
CC(CCC(=O)O)(C(=O)O)N
Quantity 100mg
Color White
Melting Point 160°C
Formula Weight 161.16
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material DL-2-Methylglutamic Acid Hemihydrate