Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2-Octanol, 97%, ACROS Organics™

Click to view available options
Quantity:
5mL
100mL
1L
2.5L
Packaging:
Glass bottle
123-96-6
C8H18O
130.231
MFCD00004591
SJWFXCIHNDVPSH-UHFFFAOYSA-N
2-octanol,2-octyl alcohol,hexylmethylcarbinol,2-hydroxyoctane,sec-caprylic alcohol,1-methylheptanol,methylhexylcarbinol,1-methylheptyl alcohol,dl-2-octanol,n-octan-2-ol
20083
CHEBI:37869
octan-2-ol
CCCCCCC(C)O
123-96-6
C8H18O
130.231
MFCD00004591
SJWFXCIHNDVPSH-UHFFFAOYSA-N
2-octanol,2-octyl alcohol,hexylmethylcarbinol,2-hydroxyoctane,sec-caprylic alcohol,1-methylheptanol,methylhexylcarbinol,1-methylheptyl alcohol,dl-2-octanol,n-octan-2-ol
20083
CHEBI:37869
octan-2-ol
CCCCCCC(C)O
Packaging Glass bottle
Color Colorless to Yellow
Quantity 5mL
Refractive Index 1.4250 to 1.4270 (20°C, 589nm)
Infrared Spectrum Authentic
Density 0.81g/mL
Assay Percent Range 97%
Linear Formula CH3(CH2)5CH(OH)CH3
Beilstein 01, 419
Fieser 01,109
Merck Index 15, 6841
Formula Weight 130.23
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material DL-2-Octanol
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger