Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2-Pentanol, 98%, Thermo Scientific Chemicals

Catalog Number AC129980025
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
6032-29-7
C5H12O
88.15
MFCD00004579
JYVLIDXNZAXMDK-UHFFFAOYSA-N
2-pentanol, sec-amyl alcohol, methylpropylcarbinol, sec-pentyl alcohol, pentanol-2, methyl propyl carbinol, 2-pentyl alcohol, 2-hydroxypentane, 1-methyl-1-butanol, alpha-methylbutanol
22386
CHEBI:77518
pentan-2-ol
CCCC(C)O
6032-29-7
C5H12O
88.15
MFCD00004579
JYVLIDXNZAXMDK-UHFFFAOYSA-N
2-pentanol, sec-amyl alcohol, methylpropylcarbinol, sec-pentyl alcohol, pentanol-2, methyl propyl carbinol, 2-pentyl alcohol, 2-hydroxypentane, 1-methyl-1-butanol, alpha-methylbutanol
22386
CHEBI:77518
pentan-2-ol
CCCC(C)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -50°C
Color Colorless
Density 0.8120g/mL
Boiling Point 118°C to 119°C
Flash Point 34°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4050 to 1.407
Linear Formula CH3CH2CH2CH(OH)CH3
Quantity 2.5 L
Residue after Evaporation 0.003% max.
Beilstein 01, 384
Specific Gravity 0.812
Merck Index 15, 7231
Solubility Information Solubility in water: very soluble. Other solubilities: miscible with alcohol and ether
Viscosity 4.1 mPa.s (20°C)
Formula Weight 88.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material DL-2-Pentanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
May cause respiratory irritation.
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Avoid breathing dust/fume/gas/mist/vapors/spray.
EINECSNumber 227-907-6

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit