Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2-Phenylpropionaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
93-53-8
C9H10O
134.18
MFCD00006973
IQVAERDLDAZARL-UHFFFAOYSA-N
2-phenylpropionaldehyde,hydratropaldehyde,hydratropic aldehyde,hyacinthal,cumene aldehyde,hydratropa aldehyde,hydrotropic aldehyde,2-phenyl-1-propanal,2-phenyl propionaldehyde,aldehyd hydratropovy
7146
2-phenylpropanal
CC(C=O)C1=CC=CC=C1
93-53-8
C9H10O
134.18
MFCD00006973
IQVAERDLDAZARL-UHFFFAOYSA-N
2-phenylpropionaldehyde,hydratropaldehyde,hydratropic aldehyde,hyacinthal,cumene aldehyde,hydratropa aldehyde,hydrotropic aldehyde,2-phenyl-1-propanal,2-phenyl propionaldehyde,aldehyd hydratropovy
7146
2-phenylpropanal
CC(C=O)C1=CC=CC=C1
Viscosity 2.16 mPa.s (20°C)
Specific Gravity 1.011
Refractive Index 1.5160 to 1.5240
Flash Point 76°C
Infrared Spectrum Authentic
Color Colorless
Quantity 100g
Density 1.0110g/mL
Melting Point -60.0°C
Packaging Glass bottle
Boiling Point 202.0°C to 205.0°C
Assay Percent Range 98%
Linear Formula CH3CH(C6H5)CHO
Beilstein 07, 305
Solubility Information Solubility in water: 3.4g/L (20°C).
Formula Weight 134.18
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material DL-2-Phenylpropionaldehyde
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning