Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2-Phenylpropionic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
492-37-5
C9H10O2
150.18
MFCD00002650
YPGCWEMNNLXISK-UHFFFAOYSA-N
2-phenylpropionic acid,hydratropic acid,dl-2-phenylpropionic acid,alpha-methylphenylacetic acid,2-phenyl-propionic acid,+/--2-phenylpropionic acid,alpha-methylbenzeneacetic acid,benzeneacetic acid, .alpha.-methyl,hydratropasaeure,alpha-phenylpropionic acid
10296
CHEBI:48526
2-phenylpropanoic acid
CC(C1=CC=CC=C1)C(=O)O
492-37-5
C9H10O2
150.18
MFCD00002650
YPGCWEMNNLXISK-UHFFFAOYSA-N
2-phenylpropionic acid,hydratropic acid,dl-2-phenylpropionic acid,alpha-methylphenylacetic acid,2-phenyl-propionic acid,+/--2-phenylpropionic acid,alpha-methylbenzeneacetic acid,benzeneacetic acid, .alpha.-methyl,hydratropasaeure,alpha-phenylpropionic acid
10296
CHEBI:48526
2-phenylpropanoic acid
CC(C1=CC=CC=C1)C(=O)O
Refractive Index 1.5215 to 1.5235
Density 1.1000g/mL
Melting Point 5°C
Boiling Point 260°C to 262°C
Flash Point 148°C
Specific Gravity 1.1
Viscosity 44 mPa.s (20°C)
Water 0.3% max.
Infrared Spectrum Authentic
Color Yellow
Quantity 100g
Packaging Glass bottle
Assay Percent Range 98%
Linear Formula CH3CH(C6H5)CO2H
Beilstein 09, 524
Solubility Information Solubility in water: 10g/L (20°C)
Formula Weight 150.18
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material DL-2-Phenylpropionic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.