Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-3-(Methylthio)-2-butanone, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
53475-15-3
C5H10OS
118.194
MFCD00008761
HFVLNCDRAMUMCC-UHFFFAOYSA-N
3-methylthio-2-butanone,2-butanone, 3-methylthio,3-methylmercapto-2-butanone,3-methylsulfanyl butan-2-one,dl-3-methylthio-2-butanone,dl-3-methylthio butanone,acmc-1auk9,2-butanone,3-methylthio,3-methylsulfanyl-2-butanone
103788
3-methylsulfanylbutan-2-one
CC(C(=O)C)SC
53475-15-3
C5H10OS
118.194
MFCD00008761
HFVLNCDRAMUMCC-UHFFFAOYSA-N
3-methylthio-2-butanone,2-butanone, 3-methylthio,3-methylmercapto-2-butanone,3-methylsulfanyl butan-2-one,dl-3-methylthio-2-butanone,dl-3-methylthio butanone,acmc-1auk9,2-butanone,3-methylthio,3-methylsulfanyl-2-butanone
103788
3-methylsulfanylbutan-2-one
CC(C(=O)C)SC
Quantity 5g
Density 0.97g/mL
Color Green to Yellow
Refractive Index 1.4700 to 1.4720 (20°C, 589nm)
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula CH3CH(SCH3)COCH3
Beilstein 01, IV, 3993
Formula Weight 118.19
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material DL-3-(Methylthio)-2-butanone, 99%
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 258-576-6