Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-3-Methylvaleric acid, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
105-43-1
C6H12O2
116.16
MFCD00002729
IGIDLTISMCAULB-UHFFFAOYSA-N
3-methylvaleric acid,3-methyl-pentanoic acid,pentanoic acid, 3-methyl,3-methyl valeric acid,dl-3-methylvaleric acid,3-methyl pentanoic acid,sec-butylacetic acid,valeric acid, 3-methyl,beta-methylvaleric acid,3-methyl-n-valeric acid
7755
3-methylpentanoic acid
CCC(C)CC(=O)O
105-43-1
C6H12O2
116.16
MFCD00002729
IGIDLTISMCAULB-UHFFFAOYSA-N
3-methylvaleric acid,3-methyl-pentanoic acid,pentanoic acid, 3-methyl,3-methyl valeric acid,dl-3-methylvaleric acid,3-methyl pentanoic acid,sec-butylacetic acid,valeric acid, 3-methyl,beta-methylvaleric acid,3-methyl-n-valeric acid
7755
3-methylpentanoic acid
CCC(C)CC(=O)O
Specific Gravity 0.926 to 0.93 (20°/20°C)
Infrared Spectrum Authentic
Refractive Index 1.4140 to 1.4180 (20°C, 589nm)
Packaging Glass bottle
Quantity 5g
Color Colorless to Yellow
Density 0.928g/mL
Assay Percent Range 97%
Linear Formula C2H5CH(CH3)CH2CO2H
Beilstein 02, 331
Formula Weight 116.16
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material DL-3-Methylvaleric acid, 97%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger