Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DL-4-Hydroxy-3-methoxyphenylglycol, piperazine salt, 90%, Tech., Acros Organics
Click to view available options
Quantity:
100mg
Specifications
Quantity | 100mg |
Assay Percent Range | 90% |
Linear Formula | (HOC6H3(OCH3)CH(OH)CHOH)·C4H10N |
Formula Weight | 454.52 |
Grade | Technical |
Chemical Name or Material | DL-4-Hydroxy-3-methoxyphenylglycol, piperazine salt, tech., 90% |