Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-5-Benzoylamino-5-isobutyl-4-oxo-1,3-dioxane, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass Bottle
51127-26-5
C15H19NO4
277.32
MFCD01863312
ZJPYVGCXULFGJA-UHFFFAOYSA-N
n-5-2-methylpropyl-4-oxo-1,3-dioxan-5-yl benzamide,dl-5-benzoylamino-5-isobutyl-4-oxo-1,3-dioxane,95,dl-5-benzoylamino-5-isobutyl-4-oxo-1,3-dioxane,dl-5-benzoylamino-5-isobutyl-4-oxo-1?3-dioxane,5-benzoylamino-5-isobutyl-4-oxo-1,3-dioxane,n-5-2-methylpropyl-4-oxo 1,3-dioxan-5-yl benzamide,n-5-2-methyl-propyl-4-oxo-1,3 dioxan-5-yl-benzamide
2733906
N-[5-(2-methylpropyl)-4-oxo-1,3-dioxan-5-yl]benzamide
CC(C)CC1(COCOC1=O)NC(=O)C2=CC=CC=C2
51127-26-5
C15H19NO4
277.32
MFCD01863312
ZJPYVGCXULFGJA-UHFFFAOYSA-N
n-5-2-methylpropyl-4-oxo-1,3-dioxan-5-yl benzamide,dl-5-benzoylamino-5-isobutyl-4-oxo-1,3-dioxane,95,dl-5-benzoylamino-5-isobutyl-4-oxo-1,3-dioxane,dl-5-benzoylamino-5-isobutyl-4-oxo-1?3-dioxane,5-benzoylamino-5-isobutyl-4-oxo-1,3-dioxane,n-5-2-methylpropyl-4-oxo 1,3-dioxan-5-yl benzamide,n-5-2-methyl-propyl-4-oxo-1,3 dioxan-5-yl-benzamide
2733906
N-[5-(2-methylpropyl)-4-oxo-1,3-dioxan-5-yl]benzamide
CC(C)CC1(COCOC1=O)NC(=O)C2=CC=CC=C2
Packaging Glass Bottle
Quantity 5g
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Formula Weight 277.32
Physical Form Fine Crystalline Powder and Chunks
Percent Purity ≥94%
Chemical Name or Material DL-5-Benzoylamino-5-isobutyl-4-oxo-1, 3-dioxane, 95%