Alfa Aesar™ DL-5-Hydroxytryptophan, 99%

Manufacturer: Alfa Aesar™ A1223703

Quantity 1g

 View more versions of this product

Catalog No. AAA1223703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 56-69-9
    Molecular Formula C11H12N2O3
    Formula Weight 220.23
    MDL Number MFCD00005651
    Beilstein 88199
    Merck Index 14,4847
    Synonym DL-2-Amino-3-[3-(5-hydroxyindole)]propionic acid
    Chemical Name or Material DL-5-Hydroxytryptophan

    Specifications

    Formula Weight 220.23
    Molecular Formula C11H12N2O3
    CAS 56-69-9
    MDL Number MFCD00005651
    Merck Index 14,4847
    Beilstein 88199
    Melting Point ≈298°C (dec.)
    Quantity 1g
    TSCA TSCA
    Synonym DL-2-Amino-3-[3-(5-hydroxyindole)]propionic acid
    RTECS Number YN7100000
    Chemical Name or Material DL-5-Hydroxytryptophan
    Assay 99%

    Description

    Applications
    It is an immediate precursor that is used to assess central serotonergic function. It was used for the treatment of insomnia.

    Solubility
    Soluble in water (4 mg/ml at 25°C), methanol.

    Notes
    Store at 4°C. Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit