Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-6-Methoxy-α-methyl-2-naphthalenemethanol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
77301-42-9
C13H14O2
202.25
MFCD01632581
OUVJWFRUESFCCY-UHFFFAOYSA-N
1-6-methoxy-2-naphthyl ethanol,1-6-methoxynaphthalen-2-yl ethanol,1-6'-methoxy-2-naphthyl ethanol,+/--1-6-methoxynaphthyl ethanol,2-1-hydroxyethyl-6-methoxynaphthalene,1-6-methoxynaphthalen-2-yl ethan-1-ol,6-methoxy-alpha-methyl-2-naphthalenemethanol,1rs-1-6-methoxy-2-naphthyl ethanol naproxen impurity k,1-6-methoxy-2-naphthyl ethan-1-ol,naproxen impurity k
575523
1-(6-methoxynaphthalen-2-yl)ethanol
CC(C1=CC2=C(C=C1)C=C(C=C2)OC)O
77301-42-9
C13H14O2
202.25
MFCD01632581
OUVJWFRUESFCCY-UHFFFAOYSA-N
1-6-methoxy-2-naphthyl ethanol,1-6-methoxynaphthalen-2-yl ethanol,1-6'-methoxy-2-naphthyl ethanol,+/--1-6-methoxynaphthyl ethanol,2-1-hydroxyethyl-6-methoxynaphthalene,1-6-methoxynaphthalen-2-yl ethan-1-ol,6-methoxy-alpha-methyl-2-naphthalenemethanol,1rs-1-6-methoxy-2-naphthyl ethanol naproxen impurity k,1-6-methoxy-2-naphthyl ethan-1-ol,naproxen impurity k
575523
1-(6-methoxynaphthalen-2-yl)ethanol
CC(C1=CC2=C(C=C1)C=C(C=C2)OC)O
Packaging Glass bottle
Infrared Spectrum Authentic
Color Beige-Yellow to White
Quantity 5g
Melting Point 111.0°C to 114.0°C
Assay Percent Range 97.5% min. (HPLC)
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,dichloromethane,ethylacetate,slightly soluble in benzene
Formula Weight 202.25
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material DL-6-Methoxy-α-methyl-2-naphthalenemethanol, 98%
Show More Show Less