Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-7-Azatryptophan Monohydrate 98.0+%, TCI America™

Click to view available options
Quantity:
100mg
7303-50-6
C10H11N3O2
205.217
MFCD00037930
SNLOIIPRZGMRAB-UHFFFAOYSA-N
2-amino-3-1h-pyrrolo 2,3-b pyridin-3-yl propanoic acid,azatryptophan,7-azatryptophan,7-aza-dl-tryptophan,dl-7-azatryptophan,7-aza-tryptophan,d,l-7-azatryptophan,alpha-amino-1h-pyrrolo 2,3-b pyridine-3-propionic acid,3-1h-pyrrolo 2,3-b pyridin-3-yl alanine
5354789
2-amino-3-(1H-pyrrolo[2,3-b]pyridin-3-yl)propanoic acid
C1=CC2=C(NC=C2CC(C(=O)O)N)N=C1
7303-50-6
C10H11N3O2
205.217
MFCD00037930
SNLOIIPRZGMRAB-UHFFFAOYSA-N
2-amino-3-1h-pyrrolo 2,3-b pyridin-3-yl propanoic acid,azatryptophan,7-azatryptophan,7-aza-dl-tryptophan,dl-7-azatryptophan,7-aza-tryptophan,d,l-7-azatryptophan,alpha-amino-1h-pyrrolo 2,3-b pyridine-3-propionic acid,3-1h-pyrrolo 2,3-b pyridin-3-yl alanine
5354789
2-amino-3-(1H-pyrrolo[2,3-b]pyridin-3-yl)propanoic acid
C1=CC2=C(NC=C2CC(C(=O)O)N)N=C1
Melting Point 259°C
Quantity 100mg
Color White-Yellow
Formula Weight 205.22
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material DL-7-Azatryptophan Monohydrate

RTECSNumber : UY8810000

TSCA : No

Recommended Storage : Freezer