Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Alanine 99%, Thermo Scientific Chemicals

Catalog Number p-52444
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
302-72-7
C3H7NO2
89.09
MFCD00064408
QNAYBMKLOCPYGJ-UHFFFAOYNA-N
dl-alanine, alanine, alanine, dl, dl-alpha-alanine, d,l-alanine, dl-2-aminopropionic acid, r,s-alanine, dl-2-aminopropanoic acid, h-dl-ala-oh, +--alanine
602
CHEBI:16449
2-aminopropanoic acid
CC(N)C(O)=O
302-72-7
C3H7NO2
89.09
MFCD00064408
QNAYBMKLOCPYGJ-UHFFFAOYNA-N
dl-alanine, alanine, alanine, dl, dl-alpha-alanine, d,l-alanine, dl-2-aminopropionic acid, r,s-alanine, dl-2-aminopropanoic acid, h-dl-ala-oh, +--alanine
602
CHEBI:16449
2-aminopropanoic acid
CC(N)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 272.0°C to 275.0°C
Ammonium 200ppm max.
Arsenic (As) 1ppm max.
pH 5.5 to 7 (2.5% soln. at 25°C)
Color White
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Assay Percent Range 99%
Infrared Spectrum Authentic
Linear Formula CH3CH(NH2)CO2H
Iron (Fe) 10ppm max.
Beilstein 04, 387
Loss on Drying 0.2% max. (105°C, 3 hrs)
Merck Index 15, 197
Other Amino Acids 0.5% max. (TLC)
Packaging Plastic Bottle
Solubility Information Solubility in water: 156g/L in water (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and organic solvents, soluble in mineral acids, soluble in ethanol 0.0087g/100g (25°C)
Sulfate 300ppm max.
Formula Weight 89.09
Transmittance 95% min. (c=10, H2O, 430nm) 1cm cell
Percent Purity 99%
Sulfated Ash 0.1% max.
Sublimation Point >190 °C
Quantity 100 g
Physical Form Crystalline Powder or Crystals
Chemical Name or Material DL-Alanine, 99%
Show More Show Less
EINECSNumber 206-126-4
RTECSNumber AY2980000
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit