Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-alpha-Lipoic Acid, ≥98% (Dry Basis), Affymetrix/USB™

Click to view available options
Quantity:
25g
50g
100g
Packaging:
Plastic bottle
1077-28-7
C8H14O2S2
206.318
MFCD00005474
AGBQKNBQESQNJD-UHFFFAOYSA-N
dl-thioctic acid,thioctic acid,alpha-lipoic acid,5-1,2-dithiolan-3-yl pentanoic acid,dl-alpha-lipoic acid,1,2-dithiolane-3-pentanoic acid,6,8-thioctic acid,thioctacid,6-thioctic acid,alpha lipoic acid
864
CHEBI:16494
5-(dithiolan-3-yl)pentanoic acid
C1CSSC1CCCCC(=O)O
1077-28-7
C8H14O2S2
206.318
MFCD00005474
AGBQKNBQESQNJD-UHFFFAOYSA-N
dl-thioctic acid,thioctic acid,alpha-lipoic acid,5-1,2-dithiolan-3-yl pentanoic acid,dl-alpha-lipoic acid,1,2-dithiolane-3-pentanoic acid,6,8-thioctic acid,thioctacid,6-thioctic acid,alpha lipoic acid
864
CHEBI:16494
5-(dithiolan-3-yl)pentanoic acid
C1CSSC1CCCCC(=O)O
Packaging Plastic bottle
Quantity 100g
Boiling Point 160°C to 165°C
Melting Point 59°C to 63°C
Beilstein 81853
Formula Weight 206.33
Physical Form Crystalline Powder
Percent Purity ≥98%
Chemical Name or Material DL-alpha-Lipoic Acid
Assay Dry basis

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 214-071-2

RTECSNumber : JP1192000

TSCA : No

Recommended Storage : Ambient temperatures