Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-AP3, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
100mg
This item is not returnable. View return policy
20263-06-3
C3H8NO5P
169.073
LBTABPSJONFLPO-UHFFFAOYSA-N
phosphonoalanine,2-amino-3-phosphonopropionic acid,dl-2-amino-3-phosphonopropionic acid,alanine, 3-phosphono,2-amino-3-phosphonopropionate,3-phosphonoalanine,2-amino-3-phosprop,3-phosphono-dl-alanine,l-ap3,2-amino-3-phosphonopropanoate
3857
CHEBI:28388
2-amino-3-phosphonopropanoic acid
C(C(C(=O)O)N)P(=O)(O)O
This item is not returnable. View return policy
20263-06-3
C3H8NO5P
169.073
LBTABPSJONFLPO-UHFFFAOYSA-N
phosphonoalanine,2-amino-3-phosphonopropionic acid,dl-2-amino-3-phosphonopropionic acid,alanine, 3-phosphono,2-amino-3-phosphonopropionate,3-phosphonoalanine,2-amino-3-phosprop,3-phosphono-dl-alanine,l-ap3,2-amino-3-phosphonopropanoate
3857
CHEBI:28388
2-amino-3-phosphonopropanoic acid
C(C(C(=O)O)N)P(=O)(O)O

Group I mGlu antagonist; also phosphoserine phosphatase inhibitor

Competitive group I metabotropic glutamate receptor antagonist and inhibitor of phosphoserine phosphatase.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 100mg
Formula Weight 169.07
Percent Purity >98%
Chemical Name or Material DL-AP3

Recommended Storage : Store at Room Temperature