Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-AP5, Tocris Bioscience™

Catalog No. p-4783424
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective NMDA antagonist

Potent NMDA antagonist.

D-isomer, L-isomer and Sodium Salt also available.

Chemical Identifiers

CAS 76326-31-3
Molecular Formula C5H12NO5P
Molecular Weight (g/mol) 197.127
InChI Key VOROEQBFPPIACJ-UHFFFAOYSA-N
Synonym 5-phosphononorvaline, 2-amino-5-phosphovaleric acid, dl-ap5, dl-2-amino-5-phosphonovaleric acid, dl-2-amino-5-phosphonopentanoic acid, 2-amino-5-phosphonovaleric acid, 2-amino-5-phosphopentanoic acid, 2-amino-5-phosphonovalerate, 2-amino-5-phosphonopentanoate, dl-apv
PubChem CID 1216
ChEBI CHEBI:138644
IUPAC Name 2-amino-5-phosphonopentanoic acid
SMILES C(CC(C(=O)O)N)CP(=O)(O)O

Specifications

Quantity 10 mg
Formula Weight 197.13
Percent Purity >98%
Chemical Name or Material DL-AP5
Recommended Storage Store at Room Temperature
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.