Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ DL-Arginine monohydrochloride monohydrate, 98+%

Manufacturer:  Alfa Aesar™ A1075806

Quantity 5g

 View more versions of this product

Catalog No. AAA1075806


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

332360-01-7
228.69 (210.67 Anhydrous)
4158960
DL-Arginine monohydrochloride monohydrate
C6H14N4O2·HCl·H2O
MFCD00151031
DL-2-Amino-5-guanidinopentanoic acid monohydrochloride monohydrate; H-DL-Arg-OH.HCl

Specifications

332360-01-7
228.69 (210.67 Anhydrous)
4158960
5g
≥98%
C6H14N4O2·HCl·H2O
MFCD00151031
DL-2-Amino-5-guanidinopentanoic acid monohydrochloride monohydrate; H-DL-Arg-OH.HCl
DL-Arginine monohydrochloride monohydrate

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit