Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-beta-Prolinol, 97+%, Thermo Scientific™

Click to view available options
Quantity:
250mg
1g
5082-74-6
C5H11NO
101.15
MFCD07773080
QOTUIIJRVXKSJU-UHFFFAOYNA-N
3-hydroxymethylpyrrolidine, 3-pyrrolidinemethanol, pyrrolidin-3-yl-methanol, pyrrolidine-3-yl-methanol, pyrrolidin-3-yl methanol, 3-pyrrolidinylmethanol, dl-beta-prolinol, rs-3-pyrrolidinemethanol, s-3-hydroxymethyl pyrrolidine, 3-pyrrolidinemethanol, 3s
13400657
(pyrrolidin-3-yl)methanol
OCC1CCNC1
5082-74-6
C5H11NO
101.15
MFCD07773080
QOTUIIJRVXKSJU-UHFFFAOYNA-N
3-hydroxymethylpyrrolidine, 3-pyrrolidinemethanol, pyrrolidin-3-yl-methanol, pyrrolidine-3-yl-methanol, pyrrolidin-3-yl methanol, 3-pyrrolidinylmethanol, dl-beta-prolinol, rs-3-pyrrolidinemethanol, s-3-hydroxymethyl pyrrolidine, 3-pyrrolidinemethanol, 3s
13400657
(pyrrolidin-3-yl)methanol
OCC1CCNC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.
Density 0.979
Boiling Point 165°C to 171°C
Quantity 250mg
Sensitivity Hygroscopic
Beilstein 1420904
Solubility Information Slightly soluble in water
Formula Weight 101.15
Percent Purity ≥97%
Chemical Name or Material DL-beta-Prolinol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold; Store under Argon

RUO – Research Use Only