Alfa Aesar™ DL-Citrulline, 97%

Manufacturer: Alfa Aesar™ A1138606

Quantity 5g

 View more versions of this product

Catalog No. AAA1138606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 627-77-0
    Molecular Formula C6H13N3O3
    Formula Weight 175.19
    MDL Number MFCD00007955
    Synonym DL-2-Amino-5-ureidovaleric acid
    Chemical Name or Material DL-Citrulline

    Specifications

    Formula Weight 175.19
    Molecular Formula C6H13N3O3
    CAS 627-77-0
    Synonym DL-2-Amino-5-ureidovaleric acid
    Quantity 5g
    MDL Number MFCD00007955
    Chemical Name or Material DL-Citrulline
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit