Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Cysteine hydrochloride, 95%, Acros Organics™

Click to view available options
Quantity:
5g
25g
10318-18-0
C3H8ClNO2S
157.612
IFQSXNOEEPCSLW-UHFFFAOYSA-N
dl-cysteine hydrochloride,2-amino-3-mercaptopropanoic acid hydrochloride,cysteine hydrochloride,+--cysteine hydrochloride,dl-cysteine hcl,h-dl-cys-oh.hcl,cysteine, hydrochloride, dl,dl-cysteine hydrochloride hydrate,dl-cysteine hydrochloride anhydrous,2-amino-3-sulfanylpropanoic acid hydrochloride
25150
2-amino-3-sulfanylpropanoic acid;hydrochloride
C(C(C(=O)O)N)S.Cl
10318-18-0
C3H8ClNO2S
157.612
IFQSXNOEEPCSLW-UHFFFAOYSA-N
dl-cysteine hydrochloride,2-amino-3-mercaptopropanoic acid hydrochloride,cysteine hydrochloride,+--cysteine hydrochloride,dl-cysteine hcl,h-dl-cys-oh.hcl,cysteine, hydrochloride, dl,dl-cysteine hydrochloride hydrate,dl-cysteine hydrochloride anhydrous,2-amino-3-sulfanylpropanoic acid hydrochloride
25150
2-amino-3-sulfanylpropanoic acid;hydrochloride
C(C(C(=O)O)N)S.Cl
Elemental Analysis 21.49 to 24.23% (C) 8.35 to 9.42% (N)
Linear Formula HSCH2CH(NH2)CO2H·HCl
Infrared Spectrum Authentic
Loss on Drying 1% max. Vacuum, room temp, 20 hours+
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and acetone
Formula Weight 157.61
Percent Purity ≥94.0% (HPLC)
Melting Point 156.0°C to 161.0°C
Quantity 5g
Chemical Name or Material DL-Cysteine hydrochloride

GHS H Statement Causes skin irritation. Causes serious eye irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.

Warning