Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals DL-Cysteine hydrochloride hydrate, 99%

Catalog Number ac227080050
Click to view available options
Quantity:
5 g
10 g
96998-61-7
C3H8ClNO2S
157.61
MFCD00151030
IFQSXNOEEPCSLW-UHFFFAOYNA-N
dl-cysteine hydrochloride monohydrate, 2-amino-3-mercaptopropanoic acid hydrochloride hydrate, dl-cystein hydrochloride hydrate, dl-cysteine hydrochloride hydrate, dl-cys hcl h2o, cysteine hydrate hydrochloride, 2-amino-3-mercaptopropanoic acid hydrate hydrochloride, 2-azanyl-3-sulfanyl-propanoic acid hydrate hydrochloride, pubchem12372, acmc-209fdf
12219647
2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
Cl.NC(CS)C(O)=O
96998-61-7
C3H8ClNO2S
157.61
MFCD00151030
IFQSXNOEEPCSLW-UHFFFAOYNA-N
dl-cysteine hydrochloride monohydrate, 2-amino-3-mercaptopropanoic acid hydrochloride hydrate, dl-cystein hydrochloride hydrate, dl-cysteine hydrochloride hydrate, dl-cys hcl h2o, cysteine hydrate hydrochloride, 2-amino-3-mercaptopropanoic acid hydrate hydrochloride, 2-azanyl-3-sulfanyl-propanoic acid hydrate hydrochloride, pubchem12372, acmc-209fdf
12219647
2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
Cl.NC(CS)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Heavy Metals (as Pb) 10ppm max.
Assay Percent Range 99%
Linear Formula HCl·xH2O
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Loss on Drying 8.5% to 12% (vacuum, P2O5, room temp., 20 hrs)
Packaging Glass Bottle
Sulfate 280ppm max.
Transmittance 98% min. (c=10, H2O, 430nm) 1cm cell
Percent Purity 98.5 to 101.5% (on dried substance)
Sulfated Ash 0.1% max.
Quantity 5 g
Physical Form Crystalline Powder or Crystals
Chemical Name or Material DL-Cysteine hydrochloride hydrate, 99%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 233-698-2
RTECSNumber HA2200000

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit