Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-DOPA, 97%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
63-84-3
C9H11NO4
197.19
WTDRDQBEARUVNC-UHFFFAOYNA-N
2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
NC(CC1=CC=C(O)C(O)=C1)C(O)=O
63-84-3
C9H11NO4
197.19
WTDRDQBEARUVNC-UHFFFAOYNA-N
2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
NC(CC1=CC=C(O)C(O)=C1)C(O)=O

D1182, 63-84-3, (OH)2C6H3CH2CH(NH2)CO2H

Spectrum™ DL-DOPA , also known as D-3,4-dihydroxyphenylalanine or dextrodopa, is biologically inactive and is similar to L-DOPA. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Packaging Amber Glass Bottle
Quantity 5 g
CAS Max % 1
Percent Purity 97%