Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Ethionine, 99%, ACROS Organics™

Click to view available options
Quantity:
2.5g
10g
50g
Packaging:
Glass bottle
67-21-0
C6H13NO2S
163.235
MFCD00063102
GGLZPLKKBSSKCX-UHFFFAOYSA-N
dl-ethionine,s-ethylhomocysteine,aethionin,s-ethyl-dl-homocysteine,2-amino-4-ethylthio butyric acid,dl-homocysteine, s-ethyl,ethionine, dl,+--ethionine,homocysteine, s-ethyl,2-amino-4-ethylthio butanoic acid
6205
CHEBI:68662
2-amino-4-ethylsulfanylbutanoic acid
CCSCCC(C(=O)O)N
67-21-0
C6H13NO2S
163.235
MFCD00063102
GGLZPLKKBSSKCX-UHFFFAOYSA-N
dl-ethionine,s-ethylhomocysteine,aethionin,s-ethyl-dl-homocysteine,2-amino-4-ethylthio butyric acid,dl-homocysteine, s-ethyl,ethionine, dl,+--ethionine,homocysteine, s-ethyl,2-amino-4-ethylthio butanoic acid
6205
CHEBI:68662
2-amino-4-ethylsulfanylbutanoic acid
CCSCCC(C(=O)O)N
Assay Percent Range 98%
Linear Formula C2H5SCH2CH2CH(NH2)CO2H
Infrared Spectrum Authentic
Merck Index 15, 3792
Packaging Glass bottle
Formula Weight 163.23
Physical Form Crystalline Powder or Flakes
Percent Purity ≥97.5%
Color White
Melting Point 265°C to 269°C
Quantity 50g
Chemical Name or Material DL-Ethionine
Show More Show Less

GHS H Statement:
Suspected of causing genetic defects.
Suspected of causing cancer.
Suspected of damaging fertility.

GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.