Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Ethyl 2-bromobutyrate, 97%, Acros Organics

Click to view available options
Quantity:
500mL
Packaging:
Glass bottle
533-68-6
C6H11BrO2
195.056
MFCD00000155
XIMFCGSNSKXPBO-UHFFFAOYSA-N
ethyl 2-bromobutyrate,dl-ethyl 2-bromobutyrate,2-bromobutanoic acid ethyl ester,butanoic acid, 2-bromo-, ethyl ester,2-bromobutyric acid ethyl ester,ethyl .alpha.-bromobutyrate,2-bromo-butyric acid ethyl ester,butyric acid, 2-bromo-, ethyl ester,2-bromo-butanoicaciethylester,ethyl alpha-bromobutyrate
79039
ethyl 2-bromobutanoate
CCC(C(=O)OCC)Br
533-68-6
C6H11BrO2
195.056
MFCD00000155
XIMFCGSNSKXPBO-UHFFFAOYSA-N
ethyl 2-bromobutyrate,dl-ethyl 2-bromobutyrate,2-bromobutanoic acid ethyl ester,butanoic acid, 2-bromo-, ethyl ester,2-bromobutyric acid ethyl ester,ethyl .alpha.-bromobutyrate,2-bromo-butyric acid ethyl ester,butyric acid, 2-bromo-, ethyl ester,2-bromo-butanoicaciethylester,ethyl alpha-bromobutyrate
79039
ethyl 2-bromobutanoate
CCC(C(=O)OCC)Br
Refractive Index 1.4460 to 1.4480 (20°C, 589nm)
Infrared Spectrum Authentic
Density 1.3200g/mL
Quantity 500mL
Color Colorless to Yellow
Packaging Glass bottle
Assay Percent Range 97%
Linear Formula C2H5CH(Br)CO2C2H5
Beilstein 02, II, 255
Merck Index 15, 1420
Formula Weight 195.06
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material DL-Ethyl 2-bromobutyrate, 97%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger