Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Ethyl 2-bromopropionate, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
250g
1kg
Packaging:
Glass bottle
535-11-5
C5H9BrO2
181.03
MFCD00000144
ARFLASKVLJTEJD-UHFFFAOYSA-N
ethyl 2-bromopropionate,ethyl alpha-bromopropionate,ethyl-2-bromopropionate,2-bromopropanoic acid ethyl ester,propanoic acid, 2-bromo-, ethyl ester,alpha-bromopropionic acid ethyl ester,ethyl a-bromopropionate,dl-ethyl 2-bromopropionate,2-bromopropionic acid ethyl ester,ethyl alpha-bromopropanoate
79040
ethyl 2-bromopropanoate
CCOC(=O)C(C)Br
535-11-5
C5H9BrO2
181.03
MFCD00000144
ARFLASKVLJTEJD-UHFFFAOYSA-N
ethyl 2-bromopropionate,ethyl alpha-bromopropionate,ethyl-2-bromopropionate,2-bromopropanoic acid ethyl ester,propanoic acid, 2-bromo-, ethyl ester,alpha-bromopropionic acid ethyl ester,ethyl a-bromopropionate,dl-ethyl 2-bromopropionate,2-bromopropionic acid ethyl ester,ethyl alpha-bromopropanoate
79040
ethyl 2-bromopropanoate
CCOC(=O)C(C)Br
Boiling Point 156°C to 160°C
Flash Point 51°C
Specific Gravity 1.394
Refractive Index 1.4450 to 1.447
Packaging Glass bottle
Infrared Spectrum Authentic
Density 1.3940g/mL
Color Colorless to Yellow
Quantity 1kg
Assay Percent Range 99%
Linear Formula CH3CH(Br)CO2C2H5
Beilstein 02,255
Merck Index 15,3827
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol and ether
Formula Weight 181.03
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material DL-Ethyl 2-bromopropionate, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 208-609-5

RTECSNumber : UA2451730

TSCA : TSCA