Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Glyceraldehyde dimer, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
23147-59-3
C6H12O6
180.16
HQAMJDIBKAKYMU-UHFFFAOYSA-N
3,6-bis hydroxymethyl-1,4-dioxane-2,5-diol,pubchem10426,dl-glyceraldehyde dimer+,2,5-dihydroxy-3,6-dihydroxymethyl-1,4-dioxane
251615
3,6-bis(hydroxymethyl)-1,4-dioxane-2,5-diol
C(C1C(OC(C(O1)O)CO)O)O
23147-59-3
C6H12O6
180.16
HQAMJDIBKAKYMU-UHFFFAOYSA-N
3,6-bis hydroxymethyl-1,4-dioxane-2,5-diol,pubchem10426,dl-glyceraldehyde dimer+,2,5-dihydroxy-3,6-dihydroxymethyl-1,4-dioxane
251615
3,6-bis(hydroxymethyl)-1,4-dioxane-2,5-diol
C(C1C(OC(C(O1)O)CO)O)O
Packaging Glass Bottle
Melting Point 138.0°C
Infrared Spectrum Authentic
Quantity 1g
Water 1% max.
Assay Percent Range 94% min. (GC)
Formula Weight 180.16
Percent Purity 95%
Chemical Name or Material DL-Glyceraldehyde dimer