Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Homocysteinethiolactone Hydrochloride 99%, ACROS Organics™

Click to view available options
Quantity:
1g
25g
100g
Packaging:
Glass bottle
6038-19-3
C4H8ClNOS
153.624
MFCD00012724
ZSEGSUBKDDEALH-UHFFFAOYSA-N
dl-homocysteine thiolactone hydrochloride,3-aminodihydrothiophen-2 3h-one hydrochloride,3-aminothiolan-2-one hydrochloride,hctl hydrochloride,dl-homocysteinethiolactone hydrochloride,d,l-homocysteine thiolactone hydrochloride,d,l-homocysteinthiolakton chlorid german,+--dihydro-3-amino-2 3h-thiophenone hydrochloride,2 3h-thiophenone, dihydro-3-amino-, hydrochloride, +-,l-2-amino-4-mercaptobutyric acid 1,4-thiolactone hydrochloride
110753
3-aminothiolan-2-one;hydrochloride
C1CSC(=O)C1N.Cl
6038-19-3
C4H8ClNOS
153.624
MFCD00012724
ZSEGSUBKDDEALH-UHFFFAOYSA-N
dl-homocysteine thiolactone hydrochloride,3-aminodihydrothiophen-2 3h-one hydrochloride,3-aminothiolan-2-one hydrochloride,hctl hydrochloride,dl-homocysteinethiolactone hydrochloride,d,l-homocysteine thiolactone hydrochloride,d,l-homocysteinthiolakton chlorid german,+--dihydro-3-amino-2 3h-thiophenone hydrochloride,2 3h-thiophenone, dihydro-3-amino-, hydrochloride, +-,l-2-amino-4-mercaptobutyric acid 1,4-thiolactone hydrochloride
110753
3-aminothiolan-2-one;hydrochloride
C1CSC(=O)C1N.Cl
Quantity 1g
Color White
Sublimation Point >140°C
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 99%
Merck Index 15, 4772
Formula Weight 153.63
Physical Form Fine Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material DL-Homocysteinethiolactone hydrochloride, 99%
Show More Show Less

EINECSNumber : 227-923-3

RTECSNumber : XN1928400