Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ DL-Homoserine, 98%

Click to view available options
Quantity:
250mg
1g
5g
1927-25-9
C4H9NO3
119.12
MFCD00002618
UKAUYVFTDYCKQA-UHFFFAOYSA-N
dl-homoserine,homoserine,h-dl-hoser-oh,d,l-homoserine,h-d-homoser-oh,butyric acid, 2-amino-4-hydroxy-, dl,2-amino-4-hydroxy-butanoic acid,dl-2-amino-4-hydroxybutyric acid,d-homoserin,h-dl-hse-oh
779
CHEBI:30653
2-amino-4-hydroxybutanoic acid
C(CO)C(C(=O)O)N
1927-25-9
C4H9NO3
119.12
MFCD00002618
UKAUYVFTDYCKQA-UHFFFAOYSA-N
dl-homoserine,homoserine,h-dl-hoser-oh,d,l-homoserine,h-d-homoser-oh,butyric acid, 2-amino-4-hydroxy-, dl,2-amino-4-hydroxy-butanoic acid,dl-2-amino-4-hydroxybutyric acid,d-homoserin,h-dl-hse-oh
779
CHEBI:30653
2-amino-4-hydroxybutanoic acid
C(CO)C(C(=O)O)N
Beilstein 1721682
Merck Index 14,4739
Formula Weight 119.12
Physical Form Solid
Percent Purity 98%
Melting Point 188°C to 189°C
Quantity 250mg
Chemical Name or Material DL-Homoserine

EINECSNumber : 217-661-8

TSCA : No

Recommended Storage : Ambient temperatures