Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Indole-3-lactic acid, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
832-97-3
C11H11NO3
205.213
MFCD00005642
XGILAAMKEQUXLS-UHFFFAOYSA-N
indole-3-lactic acid,2-hydroxy-3-1h-indol-3-yl propanoic acid,dl-indole-3-lactic acid,indolelactate,indolelactic acid,3-indolelactic acid,3-indol-3-yl lactic acid,alpha-hydroxy-1h-indole-3-propanoic acid,1-alpha-hydroxy-1h-indole-3-propionic acid,2-hydroxy-3-indol-3-ylpropanoic acid
92904
CHEBI:24813
2-hydroxy-3-(1H-indol-3-yl)propanoic acid
C1=CC=C2C(=C1)C(=CN2)CC(C(=O)O)O
832-97-3
C11H11NO3
205.213
MFCD00005642
XGILAAMKEQUXLS-UHFFFAOYSA-N
indole-3-lactic acid,2-hydroxy-3-1h-indol-3-yl propanoic acid,dl-indole-3-lactic acid,indolelactate,indolelactic acid,3-indolelactic acid,3-indol-3-yl lactic acid,alpha-hydroxy-1h-indole-3-propanoic acid,1-alpha-hydroxy-1h-indole-3-propionic acid,2-hydroxy-3-indol-3-ylpropanoic acid
92904
CHEBI:24813
2-hydroxy-3-(1H-indol-3-yl)propanoic acid
C1=CC=C2C(=C1)C(=CN2)CC(C(=O)O)O
Packaging Glass bottle
Quantity 5g
Melting Point 144°C to 147°C
Color Brown to White
Infrared Spectrum Authentic
Beilstein 22, I, 553
Formula Weight 205.21
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material DL-Indole-3-lactic acid