Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Kavain, ≥95% (HPLC), MilliporeSigma™ Supelco™

DL-Kavain, an analog of yangonin, is a kavalactone found in the kava plant.

Specifications

CAS 3155-48-4
Percent Purity ≥95% (HPLC)
Grade Analytical Standard
Linear Formula C14H14O3
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-3905
View Documents
MilliporeSigma Supelco
597805GF
5 g This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
11-101-3906
View Documents
MilliporeSigma Supelco
59780500MGF
500 mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Featured Industries: Food and Beverages
Specifications

Specifications

3155-48-4
Analytical Standard
Neat
≥95% (HPLC)
C14H14O3
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit