Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Malic acid, +99%, ACROS Organics™

Click to view available options
Quantity:
250g
500g
1kg
5kg
Packaging:
Plastic bottle
6915-15-7
C4H6O5
134.087
MFCD00064212
BJEPYKJPYRNKOW-UHFFFAOYSA-N
malic acid,dl-malic acid,2-hydroxysuccinic acid,malate,hydroxysuccinic acid,butanedioic acid, hydroxy,kyselina jablecna,pomalus acid,hydroxybutanedioic acid,deoxytetraric acid
525
CHEBI:6650
2-hydroxybutanedioic acid
C(C(C(=O)O)O)C(=O)O
6915-15-7
C4H6O5
134.087
MFCD00064212
BJEPYKJPYRNKOW-UHFFFAOYSA-N
malic acid,dl-malic acid,2-hydroxysuccinic acid,malate,hydroxysuccinic acid,butanedioic acid, hydroxy,kyselina jablecna,pomalus acid,hydroxybutanedioic acid,deoxytetraric acid
525
CHEBI:6650
2-hydroxybutanedioic acid
C(C(C(=O)O)O)C(=O)O
Assay Percent Range 99+%
Linear Formula HO2CCH2CH(OH)CO2H
Formula Weight 134.09
Percent Purity 99+%
Packaging Plastic bottle
Solubility Solubility in water: 558g/L (20°C). Other solubilities: 82.70 g/100 g methanol-17.75 g/100 g acetone (20°C, 45.53 g/100 g ethanol-20.70 g/ 100 g dioxane (20°C, 0.84 g/100 g diethyl ether (20°C), practically insoluble in benzene
Melting Point 128.0°C to 132.0°C
Quantity 250g
Flash Point 203°C
Chemical Name or Material DL-Malic acid

GHS P Statement Causes serious eye irritation.

GHS P Statement Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger