Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Malic acid, +99%, Thermo Scientific Chemicals

Catalog Number p-3757426
Click to view available options
Quantity:
250 g
500 g
1 kg
5 kg
Packaging:
Plastic Drum
Plastic bottle
6915-15-7
C4H6O5
134.087
MFCD00064212
BJEPYKJPYRNKOW-UHFFFAOYSA-N
malic acid, dl-malic acid, 2-hydroxysuccinic acid, malate, hydroxysuccinic acid, butanedioic acid, hydroxy, kyselina jablecna, pomalus acid, hydroxybutanedioic acid, deoxytetraric acid
525
CHEBI:6650
2-hydroxybutanedioic acid
C(C(C(=O)O)O)C(=O)O
6915-15-7
C4H6O5
134.087
MFCD00064212
BJEPYKJPYRNKOW-UHFFFAOYSA-N
malic acid, dl-malic acid, 2-hydroxysuccinic acid, malate, hydroxysuccinic acid, butanedioic acid, hydroxy, kyselina jablecna, pomalus acid, hydroxybutanedioic acid, deoxytetraric acid
525
CHEBI:6650
2-hydroxybutanedioic acid
C(C(C(=O)O)O)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 128.0°C to 132.0°C
Assay Percent Range 99+%
Linear Formula HO2CCH2CH(OH)CO2H
Solubility Information Solubility in water: 558g/L (20°C). Other solubilities: 82.70 g/100 g methanol-17.75 g/100 g acetone (20°C, 45.53 g/100 g ethanol-20.70 g/ 100 g dioxane (20°C, 0.84 g/100 g diethyl ether (20°C), practically insoluble in benzene
Formula Weight 134.09
Percent Purity 99+%
Quantity 250 g
Flash Point 203°C
Packaging Plastic bottle
Chemical Name or Material DL-Malic acid
Hazard Category Danger
Hazard Statement GHS P Statement Causes serious eye irritation.
Precautionary Statement GHS P Statement Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit