Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Menthol, Crystal, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
10 kg
50 kg
180 kg
Packaging:
Amber Glass Bottle
Drum
Fiber Drum
Poly Pail
89-78-1
C10H20O
156.27
NOOLISFMXDJSKH-KXUCPTDWSA-N
(1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O
89-78-1
C10H20O
156.27
NOOLISFMXDJSKH-KXUCPTDWSA-N
(1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O

ME111, 89-78-1, C10H20O

Spectrum™ DL-Menthol, Crystal, USP is used as a topical analgesic, antipruritic and decongestant. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Specific Rotation -2° to +2°
Quantity 500 g
CAS Max % 1
Percent Purity 98 to 102%