Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Mercaptosuccinic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
Plastic Bottle
70-49-5
C4H6O4S
150.15
NJRXVEJTAYWCQJ-UHFFFAOYSA-N
mercaptosuccinic acid,thiomalic acid,2-thiomalic acid,2-mercaptosuccinic acid,monomercaptosuccinic acid,butanedioic acid, mercapto,malic acid, 2-thio,mercaptosuccinate,2-sulfanylsuccinic acid,dl-mercaptosuccinic acid
6268
CHEBI:38705
2-sulfanylbutanedioic acid
C(C(C(=O)O)S)C(=O)O
70-49-5
C4H6O4S
150.15
NJRXVEJTAYWCQJ-UHFFFAOYSA-N
mercaptosuccinic acid,thiomalic acid,2-thiomalic acid,2-mercaptosuccinic acid,monomercaptosuccinic acid,butanedioic acid, mercapto,malic acid, 2-thio,mercaptosuccinate,2-sulfanylsuccinic acid,dl-mercaptosuccinic acid
6268
CHEBI:38705
2-sulfanylbutanedioic acid
C(C(C(=O)O)S)C(=O)O
Quantity 2.5kg
Color White
Melting Point 152°C to 155°C
Infrared Spectrum Authentic
Packaging Plastic Bottle
Assay Percent Range 99%
Linear Formula HO2CCH2CH(SH)CO2H
Beilstein 03,439
Merck Index 15,9496
Solubility Information Solubility in water: 150g/L (20°C). Other solubilities: 500g/L ethanol (20°C),soluble in acetone,moderately soluble in ether,chloroform,practically insoluble in benzene
Formula Weight 150.15
Physical Form Crystalline Powder
Percent Purity 98.5 to 101.5%
Chemical Name or Material DL-Mercaptosuccinic acid
Show More Show Less