Alfa Aesar™ DL-Methionine ethyl ester hydrochloride, 98+%

Manufacturer: Alfa Aesar™ H2578114

Quantity 25g

 View more versions of this product

Catalog No. AAH2578114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6297-53-6
    Molecular Formula C7H15NO2S·HCl
    Formula Weight 213.73
    MDL Number MFCD00237859
    Synonym H-DL-Met-OEt·HCl
    Chemical Name or Material DL-Methionine ethyl ester hydrochloride

    Specifications

    Formula Weight 213.73
    Melting Point 72° to 75°C
    Quantity 25g
    Molecular Formula C7H15NO2S·HCl
    MDL Number MFCD00237859
    CAS 6297-53-6
    Synonym H-DL-Met-OEt·HCl
    Recommended Storage 2° to 8°C
    Chemical Name or Material DL-Methionine ethyl ester hydrochloride
    Assay ≥98%

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit