Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ DL-Methionine ethyl ester hydrochloride, 98+%

Manufacturer:  Alfa Aesar™ H2578122

Quantity 100g

 View more versions of this product

Catalog No. AAH2578122


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

6297-53-6
213.73
H-DL-Met-OEt·HCl
C7H15NO2S·HCl
MFCD00237859
DL-Methionine ethyl ester hydrochloride

Specifications

6297-53-6
213.73
H-DL-Met-OEt·HCl
100g
≥98%
C7H15NO2S·HCl
MFCD00237859
72° to 75°C
DL-Methionine ethyl ester hydrochloride

Safety and Handling

SDS & Safety Documents

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit