Alfa Aesar™ DL-Methionine sulfone, 98%

Manufacturer: Alfa Aesar™ B2509414

Quantity 25g

 View more versions of this product

Catalog No. AAB2509414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 820-10-0
    Molecular Formula C5H11NO4S
    Formula Weight 181.21
    MDL Number MFCD00025079
    Synonym H-DL-Met(O2)-OH
    Chemical Name or Material DL-Methionine sulfone

    Specifications

    Formula Weight 181.21
    TSCA TSCA
    Molecular Formula C5H11NO4S
    MDL Number MFCD00025079
    CAS 820-10-0
    Synonym H-DL-Met(O2)-OH
    Quantity 25g
    Chemical Name or Material DL-Methionine sulfone
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit