Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ DL-Methionine sulfone, 98%

Click to view available options
Quantity:
1g
5g
25g
820-10-0
C5H11NO4S
181.206
MFCD00025079
UCUNFLYVYCGDHP-UHFFFAOYSA-N
dl-methionine sulfone,methionine sulfone,2-amino-4-methylsulfonyl butanoic acid,butanoic acid, 2-amino-4-methylsulfonyl,d,l-methionine sulfone,methioninesulfone,dl-2-amino-4-methylsulfonyl butanoic acid,2-amino-4-methylsulfonyl butyric acid,methionine, s,s-dioxide,butyric acid, 2-amino-4-methylsulfonyl-, dl
69961
2-amino-4-methylsulfonylbutanoic acid
CS(=O)(=O)CCC(C(=O)O)N
820-10-0
C5H11NO4S
181.206
MFCD00025079
UCUNFLYVYCGDHP-UHFFFAOYSA-N
dl-methionine sulfone,methionine sulfone,2-amino-4-methylsulfonyl butanoic acid,butanoic acid, 2-amino-4-methylsulfonyl,d,l-methionine sulfone,methioninesulfone,dl-2-amino-4-methylsulfonyl butanoic acid,2-amino-4-methylsulfonyl butyric acid,methionine, s,s-dioxide,butyric acid, 2-amino-4-methylsulfonyl-, dl
69961
2-amino-4-methylsulfonylbutanoic acid
CS(=O)(=O)CCC(C(=O)O)N
Formula Weight 181.21
Percent Purity 98%
Quantity 5g
Chemical Name or Material DL-Methionine sulfone