Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-α-Methylbenzylamine 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
618-36-0
C8H11N
121.18
MFCD00008069
RQEUFEKYXDPUSK-UHFFFAOYSA-N
1-phenylethylamine,alpha-methylbenzylamine,alpha-phenylethylamine,dl-alpha-methylbenzylamine,1-phenethylamine,alpha-aminoethylbenzene,1-amino-1-phenylethane,1-phenylethan-1-amine,a-methylbenzylamine,alpha-methylbenzenemethanamine
7408
CHEBI:670
1-phenylethanamine
CC(C1=CC=CC=C1)N
618-36-0
C8H11N
121.18
MFCD00008069
RQEUFEKYXDPUSK-UHFFFAOYSA-N
1-phenylethylamine,alpha-methylbenzylamine,alpha-phenylethylamine,dl-alpha-methylbenzylamine,1-phenethylamine,alpha-aminoethylbenzene,1-amino-1-phenylethane,1-phenylethan-1-amine,a-methylbenzylamine,alpha-methylbenzenemethanamine
7408
CHEBI:670
1-phenylethanamine
CC(C1=CC=CC=C1)N
Melting Point -65.0°C
Water 0.5% max. (K.F.)
Specific Gravity 0.94
Color Colorless to Yellow
Quantity 500g
Refractive Index 1.5250 to 1.5270
Flash Point 69°C
Packaging Glass bottle
Density 0.9400g/mL
Boiling Point 188.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula C6H5CH(CH3)NH2
Beilstein 12, 1094
Merck Index 14, 6026
Solubility Information Solubility in water: 4.2% (20°C). Other solubilities: miscible with alcohol and ether,soluble in most organic solvents
Formula Weight 121.18
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material DL-α-Methylbenzylamine, 99%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-545-8

TSCA : TSCA