Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-α-Methylleucine, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
144-24-1
C7H15NO2
145.202
MFCD00038403
ARSWQPLPYROOBG-UHFFFAOYSA-N
dl-alpha-methylleucine,norvaline, 2,4-dimethyl-, dl,alpha-methylleucine,dl-2-methyl leucine,usaf do-57,leucine, 2-methyl,2-methyl-dl-leucine,norvaline, 2,4-dimethyl,dl-leucine, 2-methyl,2-amino-2,4-dimethylvaleric acid
95515
2-amino-2,4-dimethylpentanoic acid
CC(C)CC(C)(C(=O)O)N
144-24-1
C7H15NO2
145.202
MFCD00038403
ARSWQPLPYROOBG-UHFFFAOYSA-N
dl-alpha-methylleucine,norvaline, 2,4-dimethyl-, dl,alpha-methylleucine,dl-2-methyl leucine,usaf do-57,leucine, 2-methyl,2-methyl-dl-leucine,norvaline, 2,4-dimethyl,dl-leucine, 2-methyl,2-amino-2,4-dimethylvaleric acid
95515
2-amino-2,4-dimethylpentanoic acid
CC(C)CC(C)(C(=O)O)N
Quantity 5g
Color White
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula (CH3)2CHCH2C(CH3)(NH2)COOH
Formula Weight 145.2
Physical Form Fluffy Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material DL-α-Methylleucine

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 205-621-2

RTECSNumber : RD2400000