Alfa Aesar™ DL-N-Acetylhomocysteine thiolactone, 99%

Manufacturer: Alfa Aesar™ B2274122

Quantity 100g

 View more versions of this product

Catalog No. AAB2274122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1195-16-0
    Molecular Formula C6H9NO2S
    Formula Weight 159.21
    MDL Number MFCD00005480
    Chemical Name or Material DL-N-Acetylhomocysteine thiolactone

    Specifications

    Formula Weight 159.21
    TSCA TSCA
    Molecular Formula C6H9NO2S
    MDL Number MFCD00005480
    CAS 1195-16-0
    RTECS Number AC8680000
    Odor Odorless
    Melting Point 109° to 111°C
    Quantity 100g
    Chemical Name or Material DL-N-Acetylhomocysteine thiolactone
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit