Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-N-Benzoyl-2-benzylserine, Acros Organics

Click to view available options
Quantity:
1g
52421-48-4
C17H17NO4
299.326
MFCD01863309
LKEOXPPWIMAHCM-UHFFFAOYSA-N
3-hydroxy-2-phenylcarbonylamino-2-benzylpropanoic acid,dl-n-benzoyl-2-benzylserine,2-benzyl-2-benzoylamino-3-hydroxypropanoic acid,2-benzyl-3-hydroxy-2-phenylformamido propanoic acid
2733903
2-benzamido-2-benzyl-3-hydroxypropanoic acid
C1=CC=C(C=C1)CC(CO)(C(=O)O)NC(=O)C2=CC=CC=C2
52421-48-4
C17H17NO4
299.326
MFCD01863309
LKEOXPPWIMAHCM-UHFFFAOYSA-N
3-hydroxy-2-phenylcarbonylamino-2-benzylpropanoic acid,dl-n-benzoyl-2-benzylserine,2-benzyl-2-benzoylamino-3-hydroxypropanoic acid,2-benzyl-3-hydroxy-2-phenylformamido propanoic acid
2733903
2-benzamido-2-benzyl-3-hydroxypropanoic acid
C1=CC=C(C=C1)CC(CO)(C(=O)O)NC(=O)C2=CC=CC=C2
Quantity 1g
Formula Weight 299.33
Chemical Name or Material DL-N-Benzoyl-2-benzylserine