Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-N-Benzoyl-2-isobutylserine, Acros Organics

Click to view available options
Quantity:
1g
5g
52421-47-3
C14H19NO4
265.309
MFCD01863308
FKUCELLBHQAFFX-UHFFFAOYSA-N
dl-n-benzoyl-2-isobutylserine,2-hydroxymethyl-4-methyl-2-phenylcarbonylamino pentanoic acid,n-benzoyl-2-hydroxymethyl leucine,2-benzamido-2-hydroxymethyl-4-methylpentanoic acid,2-hydroxymethyl-4-methyl-2-phenylformamido pentanoic acid
2733902
2-benzamido-2-(hydroxymethyl)-4-methylpentanoic acid
CC(C)CC(CO)(C(=O)O)NC(=O)C1=CC=CC=C1
52421-47-3
C14H19NO4
265.309
MFCD01863308
FKUCELLBHQAFFX-UHFFFAOYSA-N
dl-n-benzoyl-2-isobutylserine,2-hydroxymethyl-4-methyl-2-phenylcarbonylamino pentanoic acid,n-benzoyl-2-hydroxymethyl leucine,2-benzamido-2-hydroxymethyl-4-methylpentanoic acid,2-hydroxymethyl-4-methyl-2-phenylformamido pentanoic acid
2733902
2-benzamido-2-(hydroxymethyl)-4-methylpentanoic acid
CC(C)CC(CO)(C(=O)O)NC(=O)C1=CC=CC=C1
Quantity 1g
Formula Weight 265.31
Chemical Name or Material DL-N-Benzoyl-2-isobutylserine