Alfa Aesar™ DL-Norleucine, 98%

Manufacturer: Alfa Aesar™ A1079136

Quantity 500g

 View more versions of this product

Catalog No. AAA1079136

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 616-06-8
    Molecular Formula C6H13NO2
    Formula Weight 131.18
    MDL Number MFCD00064422
    Beilstein 1721748
    Merck Index 14,6706
    Synonym (+/-)-2-Aminohexanoic acid; H-DL-Nle-OH
    Chemical Name or Material DL-Norleucine

    Specifications

    Formula Weight 131.18
    TSCA TSCA
    Molecular Formula C6H13NO2
    MDL Number MFCD00064422
    Odor Odorless
    CAS 616-06-8
    Beilstein 1721748
    Merck Index 14,6706
    Synonym (+/-)-2-Aminohexanoic acid; H-DL-Nle-OH
    Melting Point >300°C
    Quantity 500g
    Chemical Name or Material DL-Norleucine
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit