Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ DL-Panthenol, 99%

Manufacturer:  Alfa Aesar™ B251140B

Quantity 1000g

 View more versions of this product

Catalog No. AAB251140B


  Check Availability
Add to cart

Specifications

Description

Applications
DL-Panthenol is used in enantiospecific HPLC separation of pantothenic acid, panthenol and pantolactone enantiomers. It promotes stimulation of epithelization, granulation, mitigation of itching and has an anti-inflammatory effect. It is effective for moisturization, improvement of hair structure and adding luster and sheen to the hair.

Solubility
Soluble in water, ethanol, ether, chloroform, and propylene glycol.

Notes
Hygroscopic. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents and moisture.
Description & Specifications

Chemical Identifiers

16485-10-2
205.26
14,2947
DL-Panthenol
C9H19NO4
MFCD00002944
DL-Pantothenol

Specifications

66° to 69°C
16485-10-2
205.26
14,2947
DL-Panthenol
1000g
C9H19NO4
MFCD00002944
DL-Pantothenol
99%

Safety and Handling

SDS & Safety Documents

RTECSNumber : ES4316500

TSCA : TSCA

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit