Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DL-Phenylalanine, ≥99% (Dry Basis), Thermo Scientific™

Click to view available options
Quantity:
100g
1kg
Packaging:
Plastic bottle
150-30-1
C9H11NO2
165.192
MFCD00064225
COLNVLDHVKWLRT-UHFFFAOYSA-N
dl-phenylalanine,phenylalanine,h-dl-phe-oh,dl-3-phenylalanine,phenylalanin,dlpa,dl-2-amino-3-phenylpropanoic acid,alanine, phenyl-, dl,2-amino-3-phenylpropionic acid,phenylalanine, dl
994
CHEBI:28044
2-amino-3-phenylpropanoic acid
C1=CC=C(C=C1)CC(C(=O)O)N
150-30-1
C9H11NO2
165.192
MFCD00064225
COLNVLDHVKWLRT-UHFFFAOYSA-N
dl-phenylalanine,phenylalanine,h-dl-phe-oh,dl-3-phenylalanine,phenylalanin,dlpa,dl-2-amino-3-phenylpropanoic acid,alanine, phenyl-, dl,2-amino-3-phenylpropionic acid,phenylalanine, dl
994
CHEBI:28044
2-amino-3-phenylpropanoic acid
C1=CC=C(C=C1)CC(C(=O)O)N
Linear Formula C6H5CH2CH(NH2)COOH
Packaging Plastic bottle
Formula Weight 165.19
Physical Form Crystalline Powder
Percent Purity ≥99%
Melting Point ~266°C (decomposition)
Quantity 1kg
Chemical Name or Material DL-Phenylalanine
Assay Dry basis