Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Phenylalanine, FCC, 98.5-101.5%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Poly Bottle
Poly Pail
150-30-1
C9H11NO2
165.19
COLNVLDHVKWLRT-UHFFFAOYNA-N
2-amino-3-phenylpropanoic acid
NC(CC1=CC=CC=C1)C(O)=O
150-30-1
C9H11NO2
165.19
COLNVLDHVKWLRT-UHFFFAOYNA-N
2-amino-3-phenylpropanoic acid
NC(CC1=CC=CC=C1)C(O)=O

P1581, 150-30-1, C9H11NO2

Spectrum™ DL-Phenylalanine, FCC is used as an amino acid nutritional supplement and flavor enhancer. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Bottle
Ignition Residue 0.003
Loss on Drying 0.002
Quantity 100 g
CAS Max % 1
Percent Purity 98.5 to 101.5%