Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Phenylsuccinic acid, 98+%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
635-51-8
C10H10O4
194.19
MFCD00004256
LVFFZQQWIZURIO-UHFFFAOYSA-N
phenylsuccinic acid,dl-phenylsuccinic acid,2-phenylsuccinic acid,phenyl succinic acid,butanedioic acid, phenyl,succinic acid, phenyl,2-phenylsuccinate,.alpha.-phenylsuccinic acid,phenylsuccinate,butanedioic acid, 2-phenyl
95459
2-phenylbutanedioic acid
C1=CC=C(C=C1)C(CC(=O)O)C(=O)O
635-51-8
C10H10O4
194.19
MFCD00004256
LVFFZQQWIZURIO-UHFFFAOYSA-N
phenylsuccinic acid,dl-phenylsuccinic acid,2-phenylsuccinic acid,phenyl succinic acid,butanedioic acid, phenyl,succinic acid, phenyl,2-phenylsuccinate,.alpha.-phenylsuccinic acid,phenylsuccinate,butanedioic acid, 2-phenyl
95459
2-phenylbutanedioic acid
C1=CC=C(C=C1)C(CC(=O)O)C(=O)O
Color Cream-White
Quantity 100g
Melting Point 166°C to 169°C
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Linear Formula HO2CCH2CH(C6H5)CO2H
Beilstein 09, 865
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in aceton and alcohols
Formula Weight 194.19
Physical Form Crystalline Powder
Percent Purity 98+%
Chemical Name or Material DL-Phenylsuccinic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-238-1