Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ DL-Pipecolinic acid, 99%
Click to view available options
Quantity:
5g
25g
100g
Description
Applications
DL-Pipecolinic acid is used as a diagnostic marker of pyridoxine-dependent epilepsy. It acts as a ligand in copper-catalyzed approach for P-arylation of organophosphorus compounds. It is used as organocatalyst and novel substrate in organic synthesis.
Solubility
Soluble in water (50 mg/ml), and boiling alcohol.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at room temperature.
DL-Pipecolinic acid is used as a diagnostic marker of pyridoxine-dependent epilepsy. It acts as a ligand in copper-catalyzed approach for P-arylation of organophosphorus compounds. It is used as organocatalyst and novel substrate in organic synthesis.
Solubility
Soluble in water (50 mg/ml), and boiling alcohol.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at room temperature.
Specifications
Melting Point | ~280°C (decomposition) |
Quantity | 25g |
Beilstein | 81095 |
Merck Index | 14,7458 |
Solubility Information | Soluble in water (50mg/ml),and boiling alcohol. |
Formula Weight | 129.16 |
Percent Purity | 99% |
Chemical Name or Material | DL-Pipecolinic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 208-616-3
RTECSNumber : TK6021000
TSCA : Yes
Recommended Storage : Ambient temperatures